Learn More
ACROBiosystems Biotinylated Human CLEC12A / MICL / CLL-1 Protein, His,Avitag™
Catalog No. 502055166
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human CLEC12A, His,Avitag (CLA-H82E6) is expressed from human 293 cells (HEK293). It contains AA His 65 - Ala 265 (Accession # Q5QGZ9-2).;This protein carries an Avi tag at the N-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 27.3 kDa. The protein migrates as 35-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human CLEC12A, His,Avitag (Cat. No. CLA-H82E6) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate, can bind Monoclonal Anti-Human CLEC12A Antibody, Human IgG1 with a linear range of 0.1-0.8 ng/mL (QC tested).;Immobilized Monoclonal Anti-Human CLEC12A Antibody, Human IgG1 at 1 μg/mL (100 μL/well) can bind Biotinylated Human CLEC12A, His,Avitag (Cat. No. CLA-H82E6) with a linear range of 5-78 ng/mL (Routinely tested).
Catalog No. 50-205-5166 Supplier ACROBiosystems Supplier No. CLAH82E625UG
Recombinant Protein;25UG;Biotinylated Human CLEC12A, His,Avitag (CLA-H82E6) is expressed from human 293 cells (HEK293). It contains AA His 65 - Ala 265 (Accession # Q5QGZ9-2).;This protein carries an Avi tag at the N-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 27.3 kDa. The protein migrates as 35-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human CLEC12A, His,Avitag (Cat. No. CLA-H82E6) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate, can bind Monoclonal Anti-Human CLEC12A Antibody, Human IgG1 with a linear range of 0.1-0.8 ng/mL (QC tested).;Immobilized Monoclonal Anti-Human CLEC12A Antibody, Human IgG1 at 1 μg/mL (100 μL/well) can bind Biotinylated Human CLEC12A, His,Avitag (Cat. No. CLA-H82E6) with a linear range of 5-78 ng/mL (Routinely tested).