Learn More
ACROBiosystems ACROBiosystems Biotinylated Human / Cynomolgus / Rhesus macaque ROR1 Protein, His,Avitag™ (MALS verified)
Catalog No. 502106709
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human / Cynomolgus / Rhesus macaque ROR1, His,Avitag (RO1-H82E6) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Glu 403 (Accession # Q01973-1). In the region Gln 30 - Glu 403, the AA sequence of Human, Cynomolgus and Rhesus macaque ROR1 are homologus.;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 45.7 kDa. The protein migrates as 66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-Human ROR1 MAb at 2 μg/mL (100 μL/well) can bind Biotinylated Human / Cynomolgus / Rhesus macaque ROR1, His,Avitag (Cat. No. RO1-H82E6) with a linear range of 0.5-4 ng/mL (QC tested).;Anti-Human ROR1 mAb captured on CM5 chip via anti-mouse antibodies surface can bind Biotinylated Human / Cynomolgus / Rhesus macaque ROR1, His, Avitag (Cat. No. RO1-H82E6) with an affinity constant of 5.81 nM as determined in a SPR assay (Biacore T200) (Routinely t
Catalog No. 50-210-6709 Supplier ACROBiosystems Supplier No. RO1H82E6200UG
Recombinant Protein;200UG;Biotinylated Human / Cynomolgus / Rhesus macaque ROR1, His,Avitag (RO1-H82E6) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Glu 403 (Accession # Q01973-1). In the region Gln 30 - Glu 403, the AA sequence of Human, Cynomolgus and Rhesus macaque ROR1 are homologus.;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 45.7 kDa. The protein migrates as 66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-Human ROR1 MAb at 2 μg/mL (100 μL/well) can bind Biotinylated Human / Cynomolgus / Rhesus macaque ROR1, His,Avitag (Cat. No. RO1-H82E6) with a linear range of 0.5-4 ng/mL (QC tested).;Anti-Human ROR1 mAb captured on CM5 chip via anti-mouse antibodies surface can bind Biotinylated Human / Cynomolgus / Rhesus macaque ROR1, His, Avitag (Cat. No. RO1-H82E6) with an affinity constant of 5.81 nM as determined in a SPR assay (Biacore T200) (Routinely t