Learn More
ACROBiosystems ACROBiosystems Biotinylated Human DPPIV / CD26 Protein, His,Avitag™
Catalog No. 502106285
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human DPPIV, His,Avitag (DP4-H82E3) is expressed from human 293 cells (HEK293). It contains AA Asp 34 - Pro 766 (Accession # NP_001926.2).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 88.4 kDa. The protein migrates as 100-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6285 Supplier ACROBiosystems Supplier No. DP4H82E325UG
Recombinant Protein;25UG;Biotinylated Human DPPIV, His,Avitag (DP4-H82E3) is expressed from human 293 cells (HEK293). It contains AA Asp 34 - Pro 766 (Accession # NP_001926.2).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 88.4 kDa. The protein migrates as 100-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.