Learn More
ACROBiosystems ACROBiosystems Biotinylated Human EpCAM / TROP1 Protein, Fc,Avitag™ (MALS verified)
Catalog No. 502106632
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human EpCAM, Fc,Avitag (EPM-H82F9) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Lys 265 (Accession # AAH14785.1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 56.2 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6632 Supplier ACROBiosystems Supplier No. EPMH82F9200UG
Recombinant Protein;200UG;Biotinylated Human EpCAM, Fc,Avitag (EPM-H82F9) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Lys 265 (Accession # AAH14785.1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 56.2 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.