Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Biotinylated Human ERBB3/HER3 protein, 72.2 kDa, >95% purity (SDS-PAGE), lyophilized from PBS, -20°C storage, C-terminal Polyhistidine tag

Catalog No. NC3654902
Encompass_Preferred

The Biotinylated Human ERBB3/HER3 protein is a high-purity recombinant protein tailored for scientific applications requiring effective biotinylation. This protein plays a significant role in studies related to ErbB3 signaling, which is crucial for cell proliferation and differentiation and is associated with several cancers. Key features include efficient labeling with AviTag™ technology and validation of bioactivity through binding assays with Human NRG1 Beta 1. This product ensures high purity and is suitable for assays involving streptavidin-based detection or immobilization.

  • Protein synonyms include ERBB3, HER3, and c-erbB3.
  • Expression system utilizes Human 293 cells (HEK293).
  • Offers a calculated molecular weight of 72.2 kDa.
  • Exhibits >95% purity as confirmed by SDS-PAGE and SEC-MALS.
  • Binding activity confirms interaction with Human NRG1 Beta 1.
  • Storage conditions include -20°C or lower for long-term storage.

Catalog No. NC3654902 Supplier ACROBiosystems Supplier No. ER3H82E6
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Biotinylated Human ERBB3/HER3 protein is a high-purity recombinant protein tailored for scientific applications requiring effective biotinylation. This protein plays a significant role in studies related to ErbB3 signaling, which is crucial for cell proliferation and differentiation and is associated with several cancers. Key features include efficient labeling with AviTag™ technology and validation of bioactivity through binding assays with Human NRG1 Beta 1. This product ensures high purity and is suitable for assays involving streptavidin-based detection or immobilization.

  • Protein synonyms include ERBB3, HER3, and c-erbB3.
  • Expression system utilizes Human 293 cells (HEK293).
  • Offers a calculated molecular weight of 72.2 kDa.
  • Exhibits >95% purity as confirmed by SDS-PAGE and SEC-MALS.
  • Binding activity confirms interaction with Human NRG1 Beta 1.
  • Storage conditions include -20°C or lower for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.