Learn More
American Research Products Inc Biotinylated human Fc gamma RI / CD64 protein 250UG
Catalog No. NC3255303
Encompass
The sequence of the extracellular domain of human CD64 (Gln 16-Leu 281) was fused with a C-terminal tag consisting of the AVI tag, TEV protease recognition sequence and a 10-His tag. The full protein sequence can be downloaded from the product webpage.
Catalog No. NC3255303 Supplier American Research Products Inc Supplier No. HUGR1B
The sequence of the extracellular domain of human CD64 (Gln 16-Leu 281) was fused with a C-terminal tag consisting of the AVI tag, TEV protease recognition sequence and a 10-His tag. The full protein sequence can be downloaded from the product webpage.