Learn More
American Research Products Inc Biotinylated human FcRn (FCGRT-B2M) heterodimer protein 250UG
Catalog No. NC3256653
Encompass
The sequence of the extracellular domain of human FCGRT (Ala 24-Ser 297) was fused with a C-terminal tag consisting of the AVI tag, TEV protease recognition sequence and a 10-His tag. This was co-transfected with the sequence of human B2M (Ile 21-Met 119) and the resulting FcRn heterodimer was purified. The full protein sequences can be downloaded from the product webpage.
Catalog No. NC3256653 Supplier American Research Products Inc Supplier No. HUFCRNB
The sequence of the extracellular domain of human FCGRT (Ala 24-Ser 297) was fused with a C-terminal tag consisting of the AVI tag, TEV protease recognition sequence and a 10-His tag. This was co-transfected with the sequence of human B2M (Ile 21-Met 119) and the resulting FcRn heterodimer was purified. The full protein sequences can be downloaded from the product webpage.