Learn More
ACROBiosystems ACROBiosystems Biotinylated Human FGF basic Protein, Avitag™,His Tag (MALS verified)
Catalog No. 502106243
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human FGF basic, Avitag,His Tag (FGC-H81E3) is expressed from E.coli cells. It contains AA Pro 143 - Ser 288 (Accession # P09038-4).;This protein carries an Avi tag at the N-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 20.1 kDa. The protein migrates as 20-22 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Human Glypican 3, Fc Tag, low endotoxin (Cat. No. GP3-H5258) at 5 μg/mL (100 μL/well) can bind Biotinylated Human FGF basic, Avitag,His Tag (Cat. No. FGC-H81E3) with a linear range of 0.6-10 ng/mL (QC tested).;Immobilized Human FGFR2 (IIIb), His Tag (Cat. No. FGB-H5223) at 5 μg/mL (100 μL/well) can bind Biotinylated Human FGF basic, Avitag,His Tag (Cat. No. FGC-H81E3) with a linear range of 0.01-0.313 μg/mL (Routinely tested).
Catalog No. 50-210-6243 Supplier ACROBiosystems Supplier No. FGCH81E325UG
Recombinant Protein;25UG;Biotinylated Human FGF basic, Avitag,His Tag (FGC-H81E3) is expressed from E.coli cells. It contains AA Pro 143 - Ser 288 (Accession # P09038-4).;This protein carries an Avi tag at the N-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 20.1 kDa. The protein migrates as 20-22 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Human Glypican 3, Fc Tag, low endotoxin (Cat. No. GP3-H5258) at 5 μg/mL (100 μL/well) can bind Biotinylated Human FGF basic, Avitag,His Tag (Cat. No. FGC-H81E3) with a linear range of 0.6-10 ng/mL (QC tested).;Immobilized Human FGFR2 (IIIb), His Tag (Cat. No. FGB-H5223) at 5 μg/mL (100 μL/well) can bind Biotinylated Human FGF basic, Avitag,His Tag (Cat. No. FGC-H81E3) with a linear range of 0.01-0.313 μg/mL (Routinely tested).