Learn More
ACROBiosystems ACROBiosystems Biotinylated Human Flt-3 / Flk-2 Protein, His,Avitag™
Catalog No. 502106899
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human Flt-3, His,Avitag™ (FL3-H82E3) is expressed from human 293 cells (HEK293). It contains AA Asn 27 - Ser 543 (Accession # P36888-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 62.1 kDa.
Catalog No. 50-210-6899 Supplier ACROBiosystems Supplier No. FL3H82E325UG
Recombinant Protein;25UG;Biotinylated Human Flt-3, His,Avitag™ (FL3-H82E3) is expressed from human 293 cells (HEK293). It contains AA Asn 27 - Ser 543 (Accession # P36888-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 62.1 kDa.