Learn More
ACROBiosystems ACROBiosystems Biotinylated Human Gastrin-17 Protein, Fc,Avitag™ (MALS verified)
Supplier:  ACROBiosystems GA7H82F425UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human Gastrin-17, Fc,Avitag (GA7-H82F4) is expressed from human 293 cells (HEK293).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 30.3 kDa. The protein migrates as 34-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-Gastrin-17 antibody, Human IgG1 at 2 μg/mL (100 μL/well) can bind Biotinylated Human Gastrin-17, Fc,Avitag (Cat. No. GA7-H82F4) with a linear range of 0.1-2.5 ng/mL (QC tested).
Catalog No. 50-210-7178