Learn More
ACROBiosystems Biotinylated Human HLA-A0201
Catalog No. 502534776
Encompass_Preferred
Biotinylated Human HLA-A*0201B2MHER2 (LIAHNQVRQV) Complex Protein (HL2-H82E3) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Ile 308 (HLA-A*0201) Ile 21 - Met 119 (B2M) LIAHNQVRQV peptide (Accession # AAA59606.1 (HLA-A*0201) P61769 (B2M) LIAHNQVRQV).
Catalog No. 50-253-4776 Supplier ACROBiosystems Supplier No. HL2H82E325UG
Biotinylated Human HLA-A*0201B2MHER2 (LIAHNQVRQV) Complex Protein (HL2-H82E3) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Ile 308 (HLA-A*0201) Ile 21 - Met 119 (B2M) LIAHNQVRQV peptide (Accession # AAA59606.1 (HLA-A*0201) P61769 (B2M) LIAHNQVRQV).