Learn More
ACROBiosystems Biotinylated Human HLA-A0201
Supplier:  ACROBiosystems HL1H82T3200UG
Encompass_Preferred
Biotinylated Human HLA-A*0201B2MNY-ESO-1 (SLLMWITQV) Complex Protein (HL1-H82T3) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Ile 308 (HLA-A*0201) Ile 21 - Met 119 (B2M) SLLMWITQV peptide (Accession # AAA59606.1 (HLA-A*0201) P61769-1 (B2M) SLLMWITQV).
Catalog No. 50-253-4785