Learn More
ACROBiosystems Biotinylated Human HLA-A0201
Supplier:  ACROBiosystems HLEH82E8200UG
Encompass_Preferred
Biotinylated Human HLA-A*0201B2MPRAME (SLLQHLIGL) Complex Protein (HLE-H82E8) is expressed from human 293 cells (HEK293). It contains AA Ile 21 - Met 119 (B2M) Gly 25 - Ile 308 (HLA-A*0201) SLLQHLIGL peptide (Accession # P61769 (B2M) AAA59606.1 (HLA-A*0201) SLLQHLIGL).
Catalog No. 50-253-4729