Learn More
ACROBiosystems Biotinylated Human HLA-A0201
Supplier:  ACROBiosystems HLCH82E525UG
Encompass_Preferred
Biotinylated Human HLA-A*0201B2MCMV pp65 (NLVPMVATV) Complex Protein (HLC-H82E5) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Ile 308 (HLA-A*0201) Ile 21 - Met 119 (B2M) NLVPMVATV peptide (Accession # AAA59606.1 (HLA-A*0201) P61769 (B2M) NLVPMVATV).
Catalog No. 50-253-4750