Learn More
ACROBiosystems Biotinylated Human HLA-A*02:01&B2M&p53 (HMTEVVRHC) Complex Protein (Monomer, MALS verified)
Catalog No. 502719407
Encompass_Preferred
TP53 has been recognized as a tumor suppressor. 50% of cancers carry a TP53 mutation while many others affect other pathway components. High-copy numbers of WT p53 peptide-MHC class I complexes were detected on tumor cells as compared to low copies on normal cells. The Human HLA-A*0201 p53 (HMTEVVRHC) complex protein is a complex of HLA-A*0201 of the MHC Class I, B2M, and HMTEVVRHC peptide of the p53.
Catalog No. 50-271-9407 Supplier ACROBiosystems Supplier No. HLPH82EF200UG
TP53 has been recognized as a tumor suppressor. 50% of cancers carry a TP53 mutation while many others affect other pathway components. High-copy numbers of WT p53 peptide-MHC class I complexes were detected on tumor cells as compared to low copies on normal cells. The Human HLA-A*0201 p53 (HMTEVVRHC) complex protein is a complex of HLA-A*0201 of the MHC Class I, B2M, and HMTEVVRHC peptide of the p53.