Learn More
ACROBiosystems Biotinylated Human HLA-A*02:01&B2M&Survivin (TLPPAWQPFL) Complex Protein (Monomer, MALS verified)
Catalog No. 502719412
Encompass_Preferred
Survivin, also known as BIRC5 and EPR-1, is a member of the inhibitor of apoptosis (IAP) family, which encodes negative regulatory proteins that prevent apoptotic cell death. IAP family members usually contain multiple baculovirus IAP repeat (BIR) domains, but birc5 encodes proteins with only a single BIR domain. The Human HLA-A*0201 Survivin (TLPPAWQPFL) complex protein is a complex of HLA-A*0201 of the MHC Class I, B2M and TLPPAWQPFL peptide of the Survivin.
Catalog No. 50-271-9412 Supplier ACROBiosystems Supplier No. HLNH82E525UG
Survivin, also known as BIRC5 and EPR-1, is a member of the inhibitor of apoptosis (IAP) family, which encodes negative regulatory proteins that prevent apoptotic cell death. IAP family members usually contain multiple baculovirus IAP repeat (BIR) domains, but birc5 encodes proteins with only a single BIR domain. The Human HLA-A*0201 Survivin (TLPPAWQPFL) complex protein is a complex of HLA-A*0201 of the MHC Class I, B2M and TLPPAWQPFL peptide of the Survivin.