Learn More
ACROBiosystems Biotinylated Human HLA-A*02:01&B2M&HPV (TIHDIILECV) Complex Protein (Monomer, MALS verified)
Catalog No. 502719382
Encompass_Preferred
Human papillomavirus (HPV) is A kind of papillomavirus belonging to the milk polypoid virus family. It is a spherical D virus, which can cause the proliferation of squamous epithelium of human skin mucosa. HPV(human papillomavirus) for common warts, genital warts (condyloma acuminatum), and other symptoms. There are many types of human papillomavirus (HPV), with HPV 16 and 18 being high-risk types known to significantly increase the risk of cervical, vaginal and vulvar cancers in women and men. The Biotin human HLA-A*0201 HPV (TIHDIILECV) protein is a complex of HLA-A*0201 of the MHC Class I, B2M, and TIHDIILECV peptide of the HPV.
Catalog No. 50-271-9382 Supplier ACROBiosystems Supplier No. HLVH82E525UG
Human papillomavirus (HPV) is A kind of papillomavirus belonging to the milk polypoid virus family. It is a spherical D virus, which can cause the proliferation of squamous epithelium of human skin mucosa. HPV(human papillomavirus) for common warts, genital warts (condyloma acuminatum), and other symptoms. There are many types of human papillomavirus (HPV), with HPV 16 and 18 being high-risk types known to significantly increase the risk of cervical, vaginal and vulvar cancers in women and men. The Biotin human HLA-A*0201 HPV (TIHDIILECV) protein is a complex of HLA-A*0201 of the MHC Class I, B2M, and TIHDIILECV peptide of the HPV.