Learn More
ACROBiosystems Biotinylated Human HLA-A0201
Supplier:  ACROBiosystems HLPH82E225UG
Encompass_Preferred
Biotinylated Human HLA-A*0201B2Mp53 (GLAPPQHLIRV) Complex Protein (HLP-H82E2) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Ile 308 (HLA-A*0201) Ile 21 - Met 119 (B2M) GLAPPQHLIRV peptide (Accession # AAA59606.1 (HLA-A*0201) P61769-1 (B2M) GLAPPQHLIRV).
Catalog No. 50-253-4698