Learn More
ACROBiosystems Biotinylated Human HLA-A0301
Catalog No. 502534752
Encompass_Preferred
Biotinylated Human HLA-A*0301B2MKRASG12C (VVVGACGVGK) Complex Protein (HLC-H82E3) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Thr 305 (HLA-A*0301) Ile 21 - Met 119 (B2M) VVVGACGVGK peptide (Accession # P04439 (HLA-A*0301) P61769 (B2M) VVVGACGVGK).
Catalog No. 50-253-4752 Supplier ACROBiosystems Supplier No. HLCH82E325UG
Biotinylated Human HLA-A*0301B2MKRASG12C (VVVGACGVGK) Complex Protein (HLC-H82E3) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Thr 305 (HLA-A*0301) Ile 21 - Met 119 (B2M) VVVGACGVGK peptide (Accession # P04439 (HLA-A*0301) P61769 (B2M) VVVGACGVGK).