Learn More
ACROBiosystems Biotinylated Human HLA-A*11:01&B2M&EBV LMP2 (SSCSSCPLSK) Complex Protein (Monomer, MALS verified)
Catalog No. 502719473
Encompass_Preferred
Epstein-Bar Virus (EBV), also known as human herpesvirus 4, belongs to gamma herpes virus family and is a very common human virus worldwide. EBV causes infectious mononucleosis (IM) and also associates to some specific types of cancers such as Burkitt’s lymphoma (BL) and gastric carcinoma (GC). Glycoprotein B (gB) plays an important role in viral entry by binding with αvβ6/αvβ8 integrins to trigger the membrane fusion and entry process of epithelial cells, which makes it become an great target for EBV research. Epstein-Bar Virus (EBV), also known as human herpesvirus 4, belongs to gamma herpes virus family and is a very common human virus worldwide. EBV causes infectious mononucleosis (IM) and also associates to some specific types of cancers such as Burkitt’s lymphoma (BL) and gastric carcinoma (GC). Glycoprotein B (gB) plays an important role in viral entry by binding with αvβ6/αvβ8 integrins to trigger the membrane fusion and entry process of epithelial cells, which makes it become
Catalog No. 50-271-9473 Supplier ACROBiosystems Supplier No. HL2H82EQ200UG
Epstein-Bar Virus (EBV), also known as human herpesvirus 4, belongs to gamma herpes virus family and is a very common human virus worldwide. EBV causes infectious mononucleosis (IM) and also associates to some specific types of cancers such as Burkitt’s lymphoma (BL) and gastric carcinoma (GC). Glycoprotein B (gB) plays an important role in viral entry by binding with αvβ6/αvβ8 integrins to trigger the membrane fusion and entry process of epithelial cells, which makes it become an great target for EBV research. Epstein-Bar Virus (EBV), also known as human herpesvirus 4, belongs to gamma herpes virus family and is a very common human virus worldwide. EBV causes infectious mononucleosis (IM) and also associates to some specific types of cancers such as Burkitt’s lymphoma (BL) and gastric carcinoma (GC). Glycoprotein B (gB) plays an important role in viral entry by binding with αvβ6/αvβ8 integrins to trigger the membrane fusion and entry process of epithelial cells, which makes it become