Learn More
ACROBiosystems Biotinylated Human HLA-A1101
Catalog No. 502534737
Encompass_Preferred
Biotinylated Human HLA-A*1101B2MKRASG12D (VVVGADGVGK) Complex Protein (HLD-H82E9) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Thr 305 (HLA-A*1101) Ile 21 - Met 119 (B2M) VVVGADGVGK peptide (Accession # Q5S3G3-1 (HLA-A*1101) P61769 (B2M) VVVGADGVGK).
Catalog No. 50-253-4737 Supplier ACROBiosystems Supplier No. HLDH82E9200UG
Biotinylated Human HLA-A*1101B2MKRASG12D (VVVGADGVGK) Complex Protein (HLD-H82E9) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Thr 305 (HLA-A*1101) Ile 21 - Met 119 (B2M) VVVGADGVGK peptide (Accession # Q5S3G3-1 (HLA-A*1101) P61769 (B2M) VVVGADGVGK).