Learn More
ACROBiosystems Biotinylated Human HLA-A*24:02&B2M&Survivin-2B (AYACNTSTL) Complex Protein (Monomer, MALS verified)
Catalog No. 502719415
Encompass_Preferred
Survivin, also known as BIRC5 and EPR-1, is a member of the inhibitor of apoptosis (IAP) family, which encodes negative regulatory proteins that prevent apoptotic cell death. IAP family members usually contain multiple baculovirus IAP repeat (BIR) domains, but birc5 encodes proteins with only a single BIR domain. The Human HLA-A*2402 Survivin-2B (AYACNTSTL) complex protein is a complex of HLA-A*2402 of the MHC Class I, B2M and AYACNTSTL peptide of the Survivin-2B.
Catalog No. 50-271-9415 Supplier ACROBiosystems Supplier No. HLNH82E3200UG
Survivin, also known as BIRC5 and EPR-1, is a member of the inhibitor of apoptosis (IAP) family, which encodes negative regulatory proteins that prevent apoptotic cell death. IAP family members usually contain multiple baculovirus IAP repeat (BIR) domains, but birc5 encodes proteins with only a single BIR domain. The Human HLA-A*2402 Survivin-2B (AYACNTSTL) complex protein is a complex of HLA-A*2402 of the MHC Class I, B2M and AYACNTSTL peptide of the Survivin-2B.