Learn More
ACROBiosystems Biotinylated Human HLA-B0702
Catalog No. 502534769
Encompass_Preferred
Biotinylated Human HLA-B*0702B2MHPV16 E7 (KPTLKEYVL) Complex Protein (HL7-H82E4) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Val 309 (HLA-B*0702) Ile 21 - Met 119 (B2M) KPTLKEYVL peptide (Accession # QDK63242.1 (HLA-B*0702) P61769 (B2M) KPTLKEYVL).
Catalog No. 50-253-4769 Supplier ACROBiosystems Supplier No. HL7H82E4200UG
Biotinylated Human HLA-B*0702B2MHPV16 E7 (KPTLKEYVL) Complex Protein (HL7-H82E4) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Val 309 (HLA-B*0702) Ile 21 - Met 119 (B2M) KPTLKEYVL peptide (Accession # QDK63242.1 (HLA-B*0702) P61769 (B2M) KPTLKEYVL).