Learn More
ACROBiosystems Biotinylated Human HLA-DRA101
Catalog No. 502534783
Encompass_Preferred
Biotinylated Human HLA-DRA1*0101HLA-DRB1*1101 Monomer Protein (HL1-H82W6) is expressed from human 293 cells (HEK293). It contains AA Ile 26 - Glu 216 (HLA-DRA1*0101) Gly 30 - Lys 227 (HLA-DRB1*1101) (Accession # CAI2388006.1 (HLA-DRA1*0101) CAI2388099.1 (HLA-DRB1*1101)).
Catalog No. 50-253-4783 Supplier ACROBiosystems Supplier No. HL1H82W6200UG
Biotinylated Human HLA-DRA1*0101HLA-DRB1*1101 Monomer Protein (HL1-H82W6) is expressed from human 293 cells (HEK293). It contains AA Ile 26 - Glu 216 (HLA-DRA1*0101) Gly 30 - Lys 227 (HLA-DRB1*1101) (Accession # CAI2388006.1 (HLA-DRA1*0101) CAI2388099.1 (HLA-DRB1*1101)).