Learn More
ACROBiosystems Biotinylated Human HLA-DRA1*01:01&HLA-DRB1*11:01&CLIP (PVSKMRMATPLLMQA) Complex Protein (Monomer)
Catalog No. 502719408
Encompass_Preferred
Biotinylated Human HLA-DRA1*01:01&HLA-DRB1*11:01&CLIP (PVSKMRMATPLLMQA) Complex Protein (HLP-H82Ed) is expressed from human 293 cells (HEK293). It contains AA Ile 26 - Glu 216 (HLA-DRA1*01:01) & Gly 30 - Lys 227 (HLA-DRB1*11:01) & PVSKMRMATPLLMQA peptide (Accession # CAI2388006.1 (HLA-DRA1*01:01) & CAI2388099.1 (HLA-DRB1*11:01) & PVSKMRMATPLLMQA).
Catalog No. 50-271-9408 Supplier ACROBiosystems Supplier No. HLPH82ED25UG
Biotinylated Human HLA-DRA1*01:01&HLA-DRB1*11:01&CLIP (PVSKMRMATPLLMQA) Complex Protein (HLP-H82Ed) is expressed from human 293 cells (HEK293). It contains AA Ile 26 - Glu 216 (HLA-DRA1*01:01) & Gly 30 - Lys 227 (HLA-DRB1*11:01) & PVSKMRMATPLLMQA peptide (Accession # CAI2388006.1 (HLA-DRA1*01:01) & CAI2388099.1 (HLA-DRB1*11:01) & PVSKMRMATPLLMQA).