Learn More
ACROBiosystems Biotinylated Human HLA-DRA1*01:01&HLA-DRB1*04:01 Monomer Protein (Peptide free, MALS verified)
Catalog No. 502719474
Encompass_Preferred
Biotinylated Human HLA-DRA1*01:01&HLA-DRB1*04:01 Monomer Protein (HL1-H82E5) is expressed from human 293 cells (HEK293). It contains AA Ile 26 - Glu 216 (HLA-DRA1*01:01) & Gly 30 - Lys 227 (HLA-DRB1*04:01) (Accession # CAI2388006.1 (HLA-DRA1*01:01) & CAI9239961.1 (HLA-DRB1*04:01)).
Catalog No. 50-271-9474 Supplier ACROBiosystems Supplier No. HL1H82E525UG
Biotinylated Human HLA-DRA1*01:01&HLA-DRB1*04:01 Monomer Protein (HL1-H82E5) is expressed from human 293 cells (HEK293). It contains AA Ile 26 - Glu 216 (HLA-DRA1*01:01) & Gly 30 - Lys 227 (HLA-DRB1*04:01) (Accession # CAI2388006.1 (HLA-DRA1*01:01) & CAI9239961.1 (HLA-DRB1*04:01)).