Learn More
ACROBiosystems Biotinylated Human HLA-DRA1*01:01&HLA-DRB1*11:01&Influenza HA (PKYVKQNTLKLAT) Complex Protein (Monomer)
Catalog No. 502719462
Encompass_Preferred
Neuraminidase (NA) and hemagglutinin (HA) are major membrane glycoproteins found on the surface of influenza virus. Hemagglutinin binds to the sialic acid-containing receptors on the surface of host cells during initial infection and at the end of an infectious cycle. Hemagglutinin also plays a major role in the determination of host range restriction and virulence. As a class I viral fusion protein, hemagglutinin is responsible for penetration of the virus into the cell cytoplasm by mediating the fusion of the membrane of the endocytosed virus particle with the endosomal membrane. The Human HLA-DRA1*0101 & HLA-DRB1*1101 Influenza HA (PKYVKQNTLKLAT) complex protein is a complex of HLA-DRA1*0101 & HLA-DRB1*1101 of the MHC Class II and PKYVKQNTLKLAT peptide of the Influenza HA.
Catalog No. 50-271-9462 Supplier ACROBiosystems Supplier No. HLAH82E725UG
Neuraminidase (NA) and hemagglutinin (HA) are major membrane glycoproteins found on the surface of influenza virus. Hemagglutinin binds to the sialic acid-containing receptors on the surface of host cells during initial infection and at the end of an infectious cycle. Hemagglutinin also plays a major role in the determination of host range restriction and virulence. As a class I viral fusion protein, hemagglutinin is responsible for penetration of the virus into the cell cytoplasm by mediating the fusion of the membrane of the endocytosed virus particle with the endosomal membrane. The Human HLA-DRA1*0101 & HLA-DRB1*1101 Influenza HA (PKYVKQNTLKLAT) complex protein is a complex of HLA-DRA1*0101 & HLA-DRB1*1101 of the MHC Class II and PKYVKQNTLKLAT peptide of the Influenza HA.