Learn More
ACROBiosystems Biotinylated Human HLA-G&B2M&Peptide (RIIPRHLQL) Complex Protein (Monomer, MALS verified)
Catalog No. 502332015
Encompass_Preferred
Biotinylated Human HLA-G&B2M&Peptide (RIIPRHLQL) Complex Protein (HLM-H82E4) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Ile 308 (HLA-G) & Ile 21 - Met 119 (B2M) & RIIPRHLQL peptide (Accession # P17693-1 (HLA-G) & P61769 (B2M) & RIIPRHLQL).
Catalog No. 50-233-2015 Supplier ACROBiosystems Supplier No. HLMH82E425UG
Biotinylated Human HLA-G&B2M&Peptide (RIIPRHLQL) Complex Protein (HLM-H82E4) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Ile 308 (HLA-G) & Ile 21 - Met 119 (B2M) & RIIPRHLQL peptide (Accession # P17693-1 (HLA-G) & P61769 (B2M) & RIIPRHLQL).