Learn More
ACROBiosystems ACROBiosystems Biotinylated Human IGF-I R / CD221 Protein, His,Avitag™
Supplier:  ACROBiosystems IGRH82E325UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human IGF-I R, His,Avitag (IGR-H82E3) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Asn 932 (Accession # P08069-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 106.6 kDa. The protein migrates as 100-130 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human IGF-I R, His,Avitag (Cat. No. IGR-H82E3) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate can bind Human IGF-I, Fc Tag (Cat. No. IG1-H4269) with a linear range of 5-156 ng/mL (QC tested).
Catalog No. 50-210-7386