Learn More
ACROBiosystems ACROBiosystems Biotinylated Human IL-17A&IL-17F Protein, Avitag™,His Tag
Catalog No. 502106680
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human IL-17A&IL-17F, Avitag,His Tag (ILF-H82W1) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Ala 155 (IL-17A) & Arg 31 - Gln 163 (IL-17F) (Accession # Q16552-1 (IL-17A) & Q96PD4-1 (IL-17F)).;Biotinylated Human IL-17A&IL-17F, Avitag,His Tag, produced by co-expression of IL-17A and IL-17F, has a calculated MW of 34.1 kDa. Subunit IL-17A&IL-17F is fused with an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The reducing (R) protein migrates as 40-43 kDa due to glycosylation.;Immobilized Anti-Human IL17A MAb at 2 μg/mL (100 μL/well) can bind Biotinylated Human IL-17A&IL-17F, Avitag,His Tag (Cat. No. ILF-H82W1) with a linear range of 0.2-6 ng/mL (QC tested).
Catalog No. 50-210-6680 Supplier ACROBiosystems Supplier No. ILFH82W125UG
Recombinant Protein;25UG;Biotinylated Human IL-17A&IL-17F, Avitag,His Tag (ILF-H82W1) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Ala 155 (IL-17A) & Arg 31 - Gln 163 (IL-17F) (Accession # Q16552-1 (IL-17A) & Q96PD4-1 (IL-17F)).;Biotinylated Human IL-17A&IL-17F, Avitag,His Tag, produced by co-expression of IL-17A and IL-17F, has a calculated MW of 34.1 kDa. Subunit IL-17A&IL-17F is fused with an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The reducing (R) protein migrates as 40-43 kDa due to glycosylation.;Immobilized Anti-Human IL17A MAb at 2 μg/mL (100 μL/well) can bind Biotinylated Human IL-17A&IL-17F, Avitag,His Tag (Cat. No. ILF-H82W1) with a linear range of 0.2-6 ng/mL (QC tested).