Learn More
ACROBiosystems ACROBiosystems Biotinylated Human IL-2 R beta&IL-2 R alpha&IL-2 R gamma Protein, Fc,Avitag™&Fc,Avitag™ (MALS verified)
Catalog No. 502106546
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human IL-2RB&IL-2RA&IL-2RG, Fc,Avitag&Fc,Avitag (ILG-H82F4) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Asp 239 & Glu 22 - Ser 212 & Leu 23 - Asn 254 (Accession # P14784-1 (IL-2RB) & P01589-1 (IL-2RA) & P31785-1 (IL-2RG)).;Biotinylated Human IL-2RB&IL-2RA&IL-2RG, Fc,Avitag&Fc,Avitag is produced by co-expression of IL-2RB and IL-2RA and IL-2RG, has a calculated MW of 52.8 kDa (IL-2RB) and 77.1 kDa (IL-2RA&IL-2RG). Subunit IL-2RB is fused with a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™) and subunit IL-2RA&IL-2RG is fused with a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The reducing (R) protein migrates as 65-70 kDa and 115-120 kDa respectively due to glycosylation.
Catalog No. 50-210-6546 Supplier ACROBiosystems Supplier No. ILGH82F4200UG
Recombinant Protein;200UG;Biotinylated Human IL-2RB&IL-2RA&IL-2RG, Fc,Avitag&Fc,Avitag (ILG-H82F4) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Asp 239 & Glu 22 - Ser 212 & Leu 23 - Asn 254 (Accession # P14784-1 (IL-2RB) & P01589-1 (IL-2RA) & P31785-1 (IL-2RG)).;Biotinylated Human IL-2RB&IL-2RA&IL-2RG, Fc,Avitag&Fc,Avitag is produced by co-expression of IL-2RB and IL-2RA and IL-2RG, has a calculated MW of 52.8 kDa (IL-2RB) and 77.1 kDa (IL-2RA&IL-2RG). Subunit IL-2RB is fused with a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™) and subunit IL-2RA&IL-2RG is fused with a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The reducing (R) protein migrates as 65-70 kDa and 115-120 kDa respectively due to glycosylation.