Recombinant Protein;25UG;Biotinylated Human IL-22, His,Avitag™ (IL2-H8247) is expressed from human 293 cells (HEK293). It contains AA Ala 34 - Ile 179 (Accession # Q9GZX6-1).;This protein carries a polyhistidine tag at the N-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 20.3 kDa. The protein migrates as 25-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human IL-22, His,Avitag (Cat. No. IL2-H8247) at 1 μg/mL (100 μL/well)on streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate. can bind Human IL-22 Antibody with a linear range of 0.1-4 ng/mL (QC tested).;Immobilized Human IL-22 Antibody at 1 μg/mL (100 μL/well) can bind Biotinylated Human IL-22, His,Avitag (Cat. No. IL2-H8247) with a linear range of 1-31 ng/mL (Routinely tested).