Learn More
ACROBiosystems Biotinylated Human IL-3 R alpha / CD123 Protein, His,Avitag™ (MALS verified)
Catalog No. 502055021
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human IL-3 R alpha, His,Avitag (ILA-H82H5) is expressed from human 293 cells (HEK293). It contains AA Thr 19 - Arg 305 (Accession # P26951-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 36.7 kDa. The protein migrates as 50-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human IL-3 R alpha, His,Avitag (Cat. No. ILA-H82H5) at 1 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate can bind Anti-IL-3 R alpha Antibody, Human IgG1 with a linear range of 0.4-2 ng/mL (QC tested).
Catalog No. 50-205-5021 Supplier ACROBiosystems Supplier No. ILAH82H5200UG
Recombinant Protein;200UG;Biotinylated Human IL-3 R alpha, His,Avitag (ILA-H82H5) is expressed from human 293 cells (HEK293). It contains AA Thr 19 - Arg 305 (Accession # P26951-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 36.7 kDa. The protein migrates as 50-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human IL-3 R alpha, His,Avitag (Cat. No. ILA-H82H5) at 1 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate can bind Anti-IL-3 R alpha Antibody, Human IgG1 with a linear range of 0.4-2 ng/mL (QC tested).