Learn More
ACROBiosystems ACROBiosystems Biotinylated Human IL-33 Protein, His,Avitag™ (MALS verified)
Catalog No. 502106532
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human IL-33, His,Avitag (IL3-H82H5) is expressed from human 293 cells (HEK293). It contains AA His 109 - Thr 270 (Accession # O95760-1).;This protein carries a polyhistidine tag at the N-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 21.9 kDa. The protein migrates as 28-33 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-1RL1, Fc Tag (Cat. No. IL1-H5250) at 5 μg/mL (100 μL/well) can bind Biotinylated Human IL-33, His,Avitag (Cat. No. IL3-H82H5) with a linear range of 0.01-0.156 μg/mL (QC tested).
Catalog No. 50-210-6532 Supplier ACROBiosystems Supplier No. IL3H82H5200UG
Recombinant Protein;200UG;Biotinylated Human IL-33, His,Avitag (IL3-H82H5) is expressed from human 293 cells (HEK293). It contains AA His 109 - Thr 270 (Accession # O95760-1).;This protein carries a polyhistidine tag at the N-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 21.9 kDa. The protein migrates as 28-33 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-1RL1, Fc Tag (Cat. No. IL1-H5250) at 5 μg/mL (100 μL/well) can bind Biotinylated Human IL-33, His,Avitag (Cat. No. IL3-H82H5) with a linear range of 0.01-0.156 μg/mL (QC tested).