Learn More
ACROBiosystems ACROBiosystems Biotinylated Human IL-4 R alpha / CD124 Protein,
Supplier:  ACROBiosystems ILRH82F425UG
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human IL-4 R alpha, (ILR-H82F4) is expressed from human 293 cells (HEK293). It contains AA Met 26 - His 232 (Accession # P24394-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 51.9 kDa.
Catalog No. 50-210-6869