Learn More
ACROBiosystems ACROBiosystems Biotinylated Human Integrin isoform alpha-7X2B beta 1 (ITGA7X2B&ITGB1) Heterodimer Protein, Tag Free&His,Avitag™
Catalog No. 502106299
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human ITGA7X2B&ITGB1 Heterodimer Protein, Tag Free&His,Avitag (IT1-H82W5) is expressed from human 293 cells (HEK293). It contains AA Phe 34 - Trp 1038 (ITGA7X2B) & Gln 21 - Asp 728 (ITGB1) (Accession # Q13683-7 (ITGA7X2B) & P05556-1 (ITGB1)).;Biotinylated Human ITGA7X2B & ITGB1 Heterodimer Protein, produced by co-expression of ITGA7X2B and ITGB1, has a calculated MW of 115.2 kDa (ITGA7X2B) and 86.7 kDa (ITGB1). Subunit ITGA7X2B is fused with an acidic tail but no tag at the C-terminus and subunit ITGB1 contains a basic tail at the C-terminus and followed by a polyhistidine tag and an Avi tag (Avitag™).
Catalog No. 50-210-6299 Supplier ACROBiosystems Supplier No. IT1H82W5200UG
Recombinant Protein;200UG;Biotinylated Human ITGA7X2B&ITGB1 Heterodimer Protein, Tag Free&His,Avitag (IT1-H82W5) is expressed from human 293 cells (HEK293). It contains AA Phe 34 - Trp 1038 (ITGA7X2B) & Gln 21 - Asp 728 (ITGB1) (Accession # Q13683-7 (ITGA7X2B) & P05556-1 (ITGB1)).;Biotinylated Human ITGA7X2B & ITGB1 Heterodimer Protein, produced by co-expression of ITGA7X2B and ITGB1, has a calculated MW of 115.2 kDa (ITGA7X2B) and 86.7 kDa (ITGB1). Subunit ITGA7X2B is fused with an acidic tail but no tag at the C-terminus and subunit ITGB1 contains a basic tail at the C-terminus and followed by a polyhistidine tag and an Avi tag (Avitag™).