Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human LILRA1 / CD85i / LIR-6 Protein, His Tag, Avi Tag (Avitag™);HEK293;25UG;LI1,LILRA1,LIR6,CD85i,LIR-6
Supplier:  ACROBiosystems LI1H82E825UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human LILRA1 / CD85i / LIR-6 Protein, His Tag, Avi Tag (Avitag™);25UG;Biotinylated Human LILRA1, His Tag, Avi Tag (LI1-H82E8) is expressed from human 293 cells (HEK293). It contains AA Pro 17 - Asn 461 (Accession # O75019-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 52.2 kDa. The protein migrates as 60-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human ANGPTL7, His Tag (Cat. No. AN7-H52H5) at 5 μg/mL (100 μL/well) can bind Biotinylated Human LILRA1, His Tag, Avi Tag (Cat. No. LI1-H82E8) with a linear range of 0.313-2.5 μg/mL (QC tested).
Catalog No. NC1461207