Learn More
ACROBiosystems ACROBiosystems Biotinylated Human LILRA2 / CD85h / ILT1 Protein, His,Avitag™
Catalog No. 502106701
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human LILRA2, His,Avitag (LI2-H82E9) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Asn 449 (Accession # Q8N149-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 50.7 kDa. The protein migrates as 67-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human ANGPTL7, His Tag (Cat. No. AN7-H52H5) at 5 μg/mL (100 μL/well) can bind Biotinylated Human LILRA2, His,Avitag (Cat. No. LI2-H82E9) with a linear range of 0.625-5 μg/mL (QC tested).
Catalog No. 50-210-6701 Supplier ACROBiosystems Supplier No. LI2H82E925UG
Recombinant Protein;25UG;Biotinylated Human LILRA2, His,Avitag (LI2-H82E9) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Asn 449 (Accession # Q8N149-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 50.7 kDa. The protein migrates as 67-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human ANGPTL7, His Tag (Cat. No. AN7-H52H5) at 5 μg/mL (100 μL/well) can bind Biotinylated Human LILRA2, His,Avitag (Cat. No. LI2-H82E9) with a linear range of 0.625-5 μg/mL (QC tested).