Learn More
ACROBiosystems ACROBiosystems Biotinylated Human LILRA3 / CD85e Protein, His,Avitag™
Catalog No. 502106704
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human LILRA3, His,Avitag (LI3-H82E0) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Glu 439 (Accession # AAH28208.1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 48.7 kDa. The protein migrates as 65-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6704 Supplier ACROBiosystems Supplier No. LI3H82E025UG
Recombinant Protein;25UG;Biotinylated Human LILRA3, His,Avitag (LI3-H82E0) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Glu 439 (Accession # AAH28208.1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 48.7 kDa. The protein migrates as 65-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.