Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human LILRA5 / CD85f / ILT11 Protein, His Tag & Avi Tag (Avitag™);HEK293;200UG;LI5,LILRA5,CD85f,ILT11,LIR9,LILRB7,ILT-11,LIR-9
Supplier:  ACROBiosystems LI5H82E1200UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human LILRA5 / CD85f / ILT11 Protein, His Tag & Avi Tag (Avitag™);200UG;Biotinylated Human LILRA5, His Tag, Avi Tag (LI5-H82E1) is expressed from human 293 cells (HEK293). It contains AA Gly 42 - Arg 268 (Accession # A6NI73-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 28.9 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human ANGPTL7, His Tag (Cat. No. AN7-H52H5) at 5 μg/mL (100 μL/well) can bind Biotinylated Human LILRA5, His Tag, Avi Tag (Cat. No. LI5-H82E1) with a linear range of 1.25-20 μg/mL (QC tested).
Catalog No. NC1477874