Learn More
ACROBiosystems ACROBiosystems Biotinylated Human LILRA5 / CD85f / ILT11 Protein, His,Avitag™
Catalog No. 502106707
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human LILRA5, His,Avitag (LI5-H82E1) is expressed from human 293 cells (HEK293). It contains AA Gly 42 - Arg 268 (Accession # A6NI73-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 28.9 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human ANGPTL7, His Tag (Cat. No. AN7-H52H5) at 5 μg/mL (100 μL/well) can bind Biotinylated Human LILRA5, His,Avitag (Cat. No. LI5-H82E1) with a linear range of 1.25-20 μg/mL (QC tested).
Catalog No. 50-210-6707 Supplier ACROBiosystems Supplier No. LI5H82E125UG
Recombinant Protein;25UG;Biotinylated Human LILRA5, His,Avitag (LI5-H82E1) is expressed from human 293 cells (HEK293). It contains AA Gly 42 - Arg 268 (Accession # A6NI73-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 28.9 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human ANGPTL7, His Tag (Cat. No. AN7-H52H5) at 5 μg/mL (100 μL/well) can bind Biotinylated Human LILRA5, His,Avitag (Cat. No. LI5-H82E1) with a linear range of 1.25-20 μg/mL (QC tested).