Learn More
ACROBiosystems ACROBiosystems Biotinylated Human LILRA6 / CD85b / ILT8 Protein, His,Avitag™
Catalog No. 502106708
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human LILRA6, His,Avitag (LI6-H82E2) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Asn 447 (Accession # Q6PI73-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 50.0 kDa. The protein migrates as 56-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6708 Supplier ACROBiosystems Supplier No. LI6H82E225UG
Recombinant Protein;25UG;Biotinylated Human LILRA6, His,Avitag (LI6-H82E2) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Asn 447 (Accession # Q6PI73-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 50.0 kDa. The protein migrates as 56-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.