Learn More
ACROBiosystems ACROBiosystems Biotinylated Human LILRB2 / CD85d / ILT4 Protein, Fc,Avitag™
Catalog No. 502106702
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human LILRB2, Fc,Avitag (LI2-H82F5) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Val 461 (Accession # AAH36827.1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 76.2 kDa. The protein migrates as 36 kDa and 100-116 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6702 Supplier ACROBiosystems Supplier No. LI2H82F5200UG
Recombinant Protein;200UG;Biotinylated Human LILRB2, Fc,Avitag (LI2-H82F5) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Val 461 (Accession # AAH36827.1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 76.2 kDa. The protein migrates as 36 kDa and 100-116 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.