Learn More
ACROBiosystems ACROBiosystems Biotinylated Human LILRB3 / CD85a / ILT5 Protein, Fc,Avitag™
Catalog No. 502106579
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human LILRB3, Fc,Avitag (CDA-H82F6) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Glu 443 (Accession # AAI12199).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 74.6 kDa. As a result of glycosylation, the protein migrates as 80-105 kDa under reducing (R) condition, and 150 kDa under non-reducing (NR) condition (SDS-PAGE).
Catalog No. 50-210-6579 Supplier ACROBiosystems Supplier No. CDAH82F6200UG
Recombinant Protein;200UG;Biotinylated Human LILRB3, Fc,Avitag (CDA-H82F6) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Glu 443 (Accession # AAI12199).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 74.6 kDa. As a result of glycosylation, the protein migrates as 80-105 kDa under reducing (R) condition, and 150 kDa under non-reducing (NR) condition (SDS-PAGE).