Learn More
ACROBiosystems ACROBiosystems Biotinylated Human LILRB4 / CD85k / ILT3 Protein, His,Avitag™ (MALS verified)
Catalog No. 502106287
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human LILRB4, His,Avitag (LI4-H82E4) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Glu 259 (Accession # AAH26309.1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 29.8 kDa. The protein migrates as 32-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6287 Supplier ACROBiosystems Supplier No. LI4H82E4200UG
Recombinant Protein;200UG;Biotinylated Human LILRB4, His,Avitag (LI4-H82E4) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Glu 259 (Accession # AAH26309.1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 29.8 kDa. The protein migrates as 32-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.