Learn More
ACROBiosystems ACROBiosystems Biotinylated Human LILRB5 / CD85c / LIR-8 Protein, Fc,Avitag™
Catalog No. 502106583
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human LILRB5, Fc,Avitag (CDC-H82F8) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Gly 458 (Accession # O75023-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 75.8 kDa. The protein migrates as 90-105 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6583 Supplier ACROBiosystems Supplier No. CDCH82F825UG
Recombinant Protein;25UG;Biotinylated Human LILRB5, Fc,Avitag (CDC-H82F8) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Gly 458 (Accession # O75023-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 75.8 kDa. The protein migrates as 90-105 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.