Learn More
ACROBiosystems ACROBiosystems Biotinylated Human LIV-1 / SLC39A6 Protein, His,Avitag™
Catalog No. 502106600
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human LIV-1, His,Avitag (LV1-H82E3) is expressed from human 293 cells (HEK293). It contains AA Phe 29 - Trp 325 (Accession # Q13433-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 37.2 kDa. The protein migrates as 55-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human LIV-1, His,Avitag (Cat. No. LV1-H82E3) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate can bind Anti-LIV-1 Antibody, Human IgG1 with a linear range of 0.6-1 ng/mL (QC tested).
Catalog No. 50-210-6600 Supplier ACROBiosystems Supplier No. LV1H82E325UG
Recombinant Protein;25UG;Biotinylated Human LIV-1, His,Avitag (LV1-H82E3) is expressed from human 293 cells (HEK293). It contains AA Phe 29 - Trp 325 (Accession # Q13433-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 37.2 kDa. The protein migrates as 55-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human LIV-1, His,Avitag (Cat. No. LV1-H82E3) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate can bind Anti-LIV-1 Antibody, Human IgG1 with a linear range of 0.6-1 ng/mL (QC tested).