Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Biotinylated human LRRC15 Fc-fusion protein (200 μg), MALS verified

Catalog No. NC2460063
Encompass_Preferred

Biotinylated human LRRC15 is a recombinant extracellular domain of LRRC15 fused to a human IgG1 Fc and a site-specific biotin acceptor tag. Supplied lyophilized for use in binding assays, ELISA, and other biochemical applications.

  • Recombinant extracellular domain fused to human IgG1 Fc and a biotin acceptor tag.
  • Expression system: HEK293 (human) cells.
  • Purity >90% by SDS-PAGE and SEC-MALS.
  • Molecular mass ~185-225 kDa by SEC-MALS; appears 95-110 kDa on SDS-PAGE due to glycosylation.
  • Amino acid range: Tyr 22-Gly 538.
  • Formulation: lyophilized with trehalose; store frozen and avoid repeated freeze-thaw.
  • Validated for binding assays and ELISA with linear detection around 0.2-10 ng/mL.

Catalog No. NC2460063 Supplier ACROBiosystems Supplier No. LR5H82F9200UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Biotinylated human LRRC15 is a recombinant extracellular domain of LRRC15 fused to a human IgG1 Fc and a site-specific biotin acceptor tag. Supplied lyophilized for use in binding assays, ELISA, and other biochemical applications.

  • Recombinant extracellular domain fused to human IgG1 Fc and a biotin acceptor tag.
  • Expression system: HEK293 (human) cells.
  • Purity >90% by SDS-PAGE and SEC-MALS.
  • Molecular mass ~185-225 kDa by SEC-MALS; appears 95-110 kDa on SDS-PAGE due to glycosylation.
  • Amino acid range: Tyr 22-Gly 538.
  • Formulation: lyophilized with trehalose; store frozen and avoid repeated freeze-thaw.
  • Validated for binding assays and ELISA with linear detection around 0.2-10 ng/mL.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.