Learn More
ACROBiosystems ACROBiosystems Biotinylated Human NCAM-1 / CD56 Protein, His,Avitag™ (MALS verified)
Catalog No. 502106611
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human NCAM-1, His,Avitag (NC1-H82E3) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Gly 718 (Accession # P13591-2).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 81.0 kDa. The protein migrates as 95-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human NCAM-1, His,Avitag (Cat. No. NC1-H82E3) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate can bind Anti-NCAM-1 Antibody, Human IgG1 with a linear range of 1-5 ng/mL (QC tested).
Catalog No. 50-210-6611 Supplier ACROBiosystems Supplier No. NC1H82E325UG
Recombinant Protein;25UG;Biotinylated Human NCAM-1, His,Avitag (NC1-H82E3) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Gly 718 (Accession # P13591-2).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 81.0 kDa. The protein migrates as 95-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human NCAM-1, His,Avitag (Cat. No. NC1-H82E3) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate can bind Anti-NCAM-1 Antibody, Human IgG1 with a linear range of 1-5 ng/mL (QC tested).