Learn More
ACROBiosystems ACROBiosystems Biotinylated Human Nectin-2 / CD112 Protein, Mouse IgG2a Fc,Avitag™ (MALS verified)
Catalog No. 502107284
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human Nectin-2, Mouse IgG2a Fc,Avitag (CD2-H82A3) is expressed from human 293 cells (HEK293). It contains AA Gln 32 - Leu 360 (Accession # Q92692-2).;This protein carries a mouse IgG2a Fc tag at the C-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 64.1 kDa. The protein migrates as 70-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human PVRIG, His Tag (Cat. No. PVG-H52H4) at 5 μg/mL (100 μL/well) can bind Biotinylated Human Nectin-2, Mouse IgG2a Fc,Avitag (Cat. No. CD2-H82A3) with a linear range of 0.6-10 ng/mL (QC tested).;Immobilized Human DNAM-1, Fc Tag (Cat. No. DN1-H5257) at 5 μg/mL (100 μL/well) can bind Biotinylated Human Nectin-2, Mouse IgG2a Fc,Avitag (Cat. No. CD2-H82A3) with a linear range of 10-78 ng/mL (Routinely tested).
Catalog No. 50-210-7284 Supplier ACROBiosystems Supplier No. CD2H82A3200UG
Recombinant Protein;200UG;Biotinylated Human Nectin-2, Mouse IgG2a Fc,Avitag (CD2-H82A3) is expressed from human 293 cells (HEK293). It contains AA Gln 32 - Leu 360 (Accession # Q92692-2).;This protein carries a mouse IgG2a Fc tag at the C-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 64.1 kDa. The protein migrates as 70-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human PVRIG, His Tag (Cat. No. PVG-H52H4) at 5 μg/mL (100 μL/well) can bind Biotinylated Human Nectin-2, Mouse IgG2a Fc,Avitag (Cat. No. CD2-H82A3) with a linear range of 0.6-10 ng/mL (QC tested).;Immobilized Human DNAM-1, Fc Tag (Cat. No. DN1-H5257) at 5 μg/mL (100 μL/well) can bind Biotinylated Human Nectin-2, Mouse IgG2a Fc,Avitag (Cat. No. CD2-H82A3) with a linear range of 10-78 ng/mL (Routinely tested).